Tisk…

Anti-MZT1 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP70912_P050
AVIVARP70912_P050EA 11290 CZK
AVIVARP70912_P050
Anti-MZT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MZT1 (mitotic spindle organizing protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR