Tisk…

Anti-KLHL33 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP70716_P050
AVIVARP70716_P050EA 11290 CZK
AVIVARP70716_P050
Anti-KLHL33 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KLHL33 (kelch-like family member 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR