Tisk…

Anti-AMMECR1L Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP68988_P050
AVIVARP68988_P050EA 11290 CZK
AVIVARP68988_P050
Anti-AMMECR1L Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AMMECR1L (AMMECR1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR