Tisk…

Anti-MOSPD1 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP67173_P050
AVIVARP67173_P050EA 11290 CZK
AVIVARP67173_P050
Anti-MOSPD1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MOSPD1 (motile sperm domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR