Tisk…

Anti-DKK3 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP63690_P050
AVIVARP63690_P050EA 11290 CZK
AVIVARP63690_P050
Anti-DKK3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DKK3 (dickkopf WNT signaling pathway inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR