Tisk…

Anti-IKBKE Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP63182_P050
AVIVARP63182_P050EA 11290 CZK
AVIVARP63182_P050
Anti-IKBKE Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR