Tisk…

Anti-DPPA4 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP57139_P050
AVIVARP57139_P050EA 11290 CZK
AVIVARP57139_P050
Anti-DPPA4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DPPA4 (developmental pluripotency associated 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR