Tisk…

Anti-DYNC1I1 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP52047_P050
AVIVARP52047_P050EA 11290 CZK
AVIVARP52047_P050
Anti-DYNC1I1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DYNC1I1 (dynein, cytoplasmic 1, intermediate chain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Guinea pig, Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR