Tisk…

Anti-CNN2 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP52039_P050
AVIVARP52039_P050EA 11290 CZK
AVIVARP52039_P050
Anti-CNN2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CNN2 (calponin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR