Tisk…

Anti-FHIT Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP51169_P050
AVIVARP51169_P050EA 11290 CZK
AVIVARP51169_P050
Anti-FHIT Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FHIT (fragile histidine triad)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR