Tisk…

Anti-ZNF226 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP50414_P050
AVIVARP50414_P050EA 11290 CZK
AVIVARP50414_P050
Anti-ZNF226 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ZNF226 (zinc finger protein 226)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR