Tisk…

Anti-ZNF223 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP35869_P050
AVIVARP35869_P050EA 11290 CZK
AVIVARP35869_P050
Anti-ZNF223 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ZNF223 (zinc finger protein 223)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR