Tisk…

Anti-HOXD12 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP32647_T100
AVIVARP32647_T100EA 9820 CZK
AVIVARP32647_T100
Anti-HOXD12 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HOXD12 (homeobox D12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR