Tisk…

Anti-POLR2B Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
ARP32457_T100
AVIVARP32457_T100EA 9820 CZK
AVIVARP32457_T100
Anti-POLR2B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: POLR2B (polymerase (RNA) II (DNA directed) polypeptide B, 140kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR