Tisk…
Dip slides
Dodavatel: PanReac AppliChem
435895.0922 435896.0922 435897.0922
APPC435895.0922EA 2390 CZK
APPC435895.0922 APPC435896.0922 APPC435897.0922
Dip slides
Média mikrobiologická Připravená mikrobiologická média
Dip slides are suitable for the microbiological hygiene control applications in both the food industry and other industrial applications.
  • Easy to use
  • Plate count ager for total aerobe count
  • Rose bengal agar for moulds and yeasts count
  • VRBG Agar for enterobacteria count
Each dip slide has two faces coated with agar-based medium, with a usable area of 12 cm² per face. Flexibility of the plastic strip allows samples to be taken in areas that are inaccessible to contact plates.
Supplied in a cardboard box and are protected by a stamped support and a sponge pad to absorb knocks and reduce the risk of the medium becoming detached.