Tisk…

4-chlor-m-krezol 99.0-101.0% NF, USP, BP, Ph. Eur., pharma grade

Dodavatel: PanReac AppliChem
Nebezpečí

Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol

145226.1211
APPC145226.1211EA 44090 CZK
APPC145226.1211
4-chlor-m-krezol 99.0-101.0% NF, USP, BP, Ph. Eur., pharma grade
4-chlor-m-krezol
Vzorec: C₇H₇ClO
Bod varu: 235 °C
Bod tání: 64…66 °C
MDL Number: MFCD00002323
CAS číslo: 59-50-7
EINECS: 200-431-6
UN: 3437
ADR: 6.1,II

Objednat

Výsledky specifikačního testu

Assay 99,0-101,0%
Identity (According to pharmacopoeias) Passes test
Acidity Passes test
Insoluble matter in CH₃CH₂OH Passes test
Non-volatile matter 0,1%
Residual solvents (Ph. Eur./USP) Passes test
Related substances (G.C.) individual 0,10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR