Tisk…
Terrestrial (Cypripedium) orchid medium, contains 400 mg/L calcium nitrate
Dodavatel: Apollo Scientific
PMT849A-10X1L PMT849B-10L PMT849C-50L
APOSPMT849A-10X1LEA 0 CZK
APOSPMT849A-10X1L APOSPMT849B-10L APOSPMT849C-50L
Terrestrial (Cypripedium) orchid medium, contains 400 mg/L calcium nitrate
Média kultivační
Contains 400 mg/L calcium nitrate, without casein. Mother flasking medium V.
  • Plant tissue culture tested