Tisk…

2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol

Dodavatel: Apollo Scientific
Varování

PC9888-50MG
APOSPC9888-50MGEA 622 CZK
APOSPC9888-50MG
2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol
2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol
Vzorec: C₁₀H₆F₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD00139776
CAS číslo: 338404-77-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR