Tisk…

1,2-Dibromo-4-fluorobenzene 98%

Dodavatel: Apollo Scientific
Varování

PC9619-100G
APOSPC9619-100GEA 5060 CZK
APOSPC9619-100G
1,2-Dibromo-4-fluorobenzene 98%
1,2-Dibromo-4-fluorobenzene
Vzorec: C₆H₃Br₂F
Storage Temperature: Ambient
MDL Number: MFCD00070780
CAS číslo: 2369-37-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR