Tisk…

6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%

Dodavatel: Apollo Scientific

PC9474-1G PC9474-500MG PC9474-5G
APOSPC9474-1GEA 4860 CZK
APOSPC9474-1G APOSPC9474-500MG APOSPC9474-5G
6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%
6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
Vzorec: C₁₄H₁₀F₃NO₃
Bod tání: 204 °C
Storage Temperature: Ambient
MDL Number: MFCD00139896
CAS číslo: 338783-19-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR