Tisk…

2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulphanyl)acetic acid

Dodavatel: Apollo Scientific

PC9469-1G PC9469-500MG PC9469-5G
APOSPC9469-1GEA 4400 CZK
APOSPC9469-1G APOSPC9469-500MG APOSPC9469-5G
2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulphanyl)acetic acid
2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulphanyl)acetic acid
Vzorec: C₁₁H₁₀F₃NO₃S
Bod tání: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00202305
CAS číslo: 338421-05-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR