Tisk…

(2,6-Difluorophenyl)acetyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC9335-1G PC9335-5G PC9335-25G
APOSPC9335-1GEA 2610 CZK
APOSPC9335-1G APOSPC9335-5G APOSPC9335-25G
(2,6-Difluorophenyl)acetyl chloride
(2,6-Difluorophenyl)acetyl chloride
Vzorec: C₈H₅ClF₂O
MW: 190.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424718
CAS číslo: 116622-90-3
UN: 1760
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR