Tisk…

2-Chloro-5-fluoropyridin-4-ol 96%

Dodavatel: Apollo Scientific

PC910219-1G PC910219-5G
APOSPC910219-1GEA 2110 CZK
APOSPC910219-1G APOSPC910219-5G
2-Chloro-5-fluoropyridin-4-ol 96%
2-Chloro-5-fluoropyridin-4-ol
Vzorec: C₅H₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD13189882
CAS číslo: 1196153-96-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR