Tisk…

2-(3-Fluorophenoxy)ethylamine hydrochloride 98%

Dodavatel: Apollo Scientific

PC908941-1G PC908941-5G
APOSPC908941-1GEA 5390 CZK
APOSPC908941-1G APOSPC908941-5G
2-(3-Fluorophenoxy)ethylamine hydrochloride 98%
2-(3-Fluorophenoxy)ethylamine hydrochloride
Vzorec: C₈H₁₁ClFNO
Storage Temperature: Ambient
MDL Number: MFCD11099553
CAS číslo: 120351-93-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR