Tisk…

N-(2,3-Difluorobenzyl)-N-methylamine 95%

Dodavatel: Apollo Scientific

PC908829-1G PC908829-5G
APOSPC908829-1GEA 8070 CZK
APOSPC908829-1G APOSPC908829-5G
N-(2,3-Difluorobenzyl)-N-methylamine 95%
N-(2,3-Difluorobenzyl)-N-methylamine
Vzorec: C₈H₉F₂N
Storage Temperature: Ambient
MDL Number: MFCD12147620
CAS číslo: 906645-41-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR