Tisk…

2,4-Difluoro-6-hydroxybenzonitrile 95%

Dodavatel: Apollo Scientific

PC908020-1G PC908020-250MG
APOSPC908020-1GEA 22950 CZK
APOSPC908020-1G APOSPC908020-250MG
2,4-Difluoro-6-hydroxybenzonitrile 95%
2,4-Difluoro-6-hydroxybenzonitrile
Vzorec: C₇H₃F₂NO
Storage Temperature: Ambient
MDL Number: MFCD18394850
CAS číslo: 1261793-35-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR