Tisk…

Ethyl 4-Fluorocinnamate 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Fluorocinnamic acid ethyl ester, Ethyl (2E)-3-(4-fluorophenyl)acrylate

PC907942-1G PC907942-5G
APOSPC907942-1GEA 4370 CZK
APOSPC907942-1G APOSPC907942-5G
Ethyl 4-Fluorocinnamate 95%
Ethyl 4-Fluorocinnamate
Vzorec: C₁₁H₁₁FO₂
MW: 194.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02252626
CAS číslo: 352-03-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR