Tisk…

3-Fluoro-N-hydroxybenzimidoyl chloride 95%

Dodavatel: Apollo Scientific

PC907276-1G
APOSPC907276-1GEA 6910 CZK
APOSPC907276-1G
3-Fluoro-N-hydroxybenzimidoyl chloride 95%
3-Fluoro-N-hydroxybenzimidoyl chloride
Vzorec: C₇H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD09831990
CAS číslo: 58606-42-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR