Tisk…

(3-Fluorocyclobutyl)methanamine hydrochloride

Dodavatel: Apollo Scientific

PC907134-1G
APOSPC907134-1GEA 39020 CZK
APOSPC907134-1G
(3-Fluorocyclobutyl)methanamine hydrochloride
(3-Fluorocyclobutyl)methanamine hydrochloride
Vzorec: C₅H₁₀FN·ClH
MW: 139.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22570249
CAS číslo: 1523606-29-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR