Tisk…

2,4,5-Trifluoro-3-ethoxybenzoic acid 95%

Dodavatel: Apollo Scientific

PC907097-1G PC907097-5G
APOSPC907097-1GEA 8800 CZK
APOSPC907097-1G APOSPC907097-5G
2,4,5-Trifluoro-3-ethoxybenzoic acid 95%
2,4,5-Trifluoro-3-ethoxybenzoic acid
Vzorec: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD08459286
CAS číslo: 169507-61-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR