Tisk…

4-Bromo-5-fluoro-2-methoxyphenol 95%

Dodavatel: Apollo Scientific

PC907036-10G PC907036-5G
APOSPC907036-10GEA 20170 CZK
APOSPC907036-10G APOSPC907036-5G
4-Bromo-5-fluoro-2-methoxyphenol 95%
4-Bromo-5-fluoro-2-methoxyphenol
Vzorec: C₇H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD08437599
CAS číslo: 886510-25-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR