Tisk…

3,5-Dimethoxy-2-fluoroaniline 95%

Dodavatel: Apollo Scientific

PC906804-1G
APOSPC906804-1GEA 18420 CZK
APOSPC906804-1G
3,5-Dimethoxy-2-fluoroaniline 95%
3,5-Dimethoxy-2-fluoroaniline
Vzorec: C₈H₁₀FNO₂
MW: 171.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20696760
CAS číslo: 651734-61-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR