Tisk…

1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

PC905485-1G
APOSPC905485-1GEA 3770 CZK
APOSPC905485-1G
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid 95%
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Vzorec: C₆H₅F₃N₂O₂
MW: 194,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04110182
CAS číslo: 119083-00-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR