Tisk…

Methyl-5-amino-2,4-difluorobenzoate 98%

Dodavatel: Apollo Scientific

Synonyms: Methyl 5-amino-2,4-difluorobenzoate

PC902802-25G
APOSPC902802-25GEA 5630 CZK
APOSPC902802-25G
Methyl-5-amino-2,4-difluorobenzoate 98%
Methyl-5-amino-2,4-difluorobenzoate
Vzorec: C₈H₇F₂NO₂
MW: 187.1435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09396710
CAS číslo: 125568-73-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR