Tisk…

2,3-Difluoropyridine-5-carboxylic acid 97%

Dodavatel: Apollo Scientific

PC901563-1G
APOSPC901563-1GEA 11540 CZK
APOSPC901563-1G
2,3-Difluoropyridine-5-carboxylic acid 97%
2,3-Difluoropyridine-5-carboxylic acid
Vzorec: C₆H₃F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD09031171
CAS číslo: 851386-33-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR