Tisk…

8-Fluoro-2-quinolinol 98%

Dodavatel: Apollo Scientific

PC900635-1G PC900635-5G
APOSPC900635-1GEA 1700 CZK
APOSPC900635-1G APOSPC900635-5G
8-Fluoro-2-quinolinol 98%
8-Fluoro-2-quinolinol
Vzorec: C₉H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD12026113
CAS číslo: 71738-83-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR