Tisk…

5-Fluoro-2-(2-methoxyethoxy)pyridine 98%

Dodavatel: Apollo Scientific

PC900589-1G PC900589-5G
APOSPC900589-1GEA 6620 CZK
APOSPC900589-1G APOSPC900589-5G
5-Fluoro-2-(2-methoxyethoxy)pyridine 98%
5-Fluoro-2-(2-methoxyethoxy)pyridine
Vzorec: C₈H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD18914536
CAS číslo: 1305322-91-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR