Tisk…

4-Fluoro-2-isopropoxyaniline 98%

Dodavatel: Apollo Scientific

PC900142-1G PC900142-5G
APOSPC900142-1GEA 7050 CZK
APOSPC900142-1G APOSPC900142-5G
4-Fluoro-2-isopropoxyaniline 98%
4-Fluoro-2-isopropoxyaniline
Vzorec: C₉H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD09738072
CAS číslo: 148583-65-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR