Tisk…

2-Fluoro-4-pyridineboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PC8520-1G PC8520-5G
APOSPC8520-1GEA 827 CZK
APOSPC8520-1G APOSPC8520-5G
2-Fluoro-4-pyridineboronic acid pinacol ester
2-Fluoro-4-pyridineboronic acid pinacol ester
Vzorec: C₁₁H₁₅BFNO₂
MW: 223.05
Bod tání: 47…51 °C
Storage Temperature: Ambient
MDL Number: MFCD06798253
CAS číslo: 458532-86-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR