Tisk…

Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate 97%

Dodavatel: Apollo Scientific

Synonyms: Pentafluorophenyl-2-(tetrahydropyran-4-yloxy)pyridine-5-carboxylate

PC8512-1G PC8512-250MG
APOSPC8512-1GEA 5230 CZK
APOSPC8512-1G APOSPC8512-250MG
Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate 97%
Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate
Vzorec: C₁₇H₁₂F₅NO₄
MW: 389.273 g/mol
Bod tání: 114.5…118 °C
Storage Temperature: Ambient
MDL Number: MFCD09702389
CAS číslo: 910036-96-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR