Tisk…

2-Bromo-4'-fluoro-2'-(trifluoromethyl)acetophenone

Dodavatel: Apollo Scientific
Nebezpečí

PC8409-1G PC8409-250MG PC8409-5G
APOSPC8409-1GEA 3970 CZK
APOSPC8409-1G APOSPC8409-250MG APOSPC8409-5G
2-Bromo-4'-fluoro-2'-(trifluoromethyl)acetophenone
2-Bromo-4'-fluoro-2'-(trifluoromethyl)acetophenone
Vzorec: C₉H₅BrF₄O
Bod varu: 62…64 °C/0,06mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD03094508
CAS číslo: 537050-13-8
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR