Tisk…

2-Bromo-2'-fluoro-3'-(trifluoromethyl)acetophenone

Dodavatel: Apollo Scientific
Nebezpečí

PC8405-1G PC8405-25G PC8405-5G
APOSPC8405-1GEA 1660 CZK
APOSPC8405-1G APOSPC8405-25G APOSPC8405-5G
2-Bromo-2'-fluoro-3'-(trifluoromethyl)acetophenone
2-Bromo-2'-fluoro-3'-(trifluoromethyl)acetophenone
Vzorec: C₉H₅BrF₄O
Storage Temperature: Refrigerator
MDL Number: MFCD03094504
CAS číslo: 537050-11-6
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR