Tisk…

Bromo(difluoro)acetyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC8341-1G PC8341-25G PC8341-5G
APOSPC8341-1GEA 2070 CZK
APOSPC8341-1G APOSPC8341-25G APOSPC8341-5G
Bromo(difluoro)acetyl bromide 97%
Bromo(difluoro)acetyl bromide
Vzorec: C₂Br₂F₂O
Bod varu: 70…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153688
CAS číslo: 1796-12-9
UN: 3265
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR