Tisk…

Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV) 98%

Dodavatel: Apollo Scientific

PC8050-1G
APOSPC8050-1GEA 3110 CZK
APOSPC8050-1G
Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV) 98%
Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV)
Vzorec: C₂₀H₁₆F₁₂O₈Zr
Bod tání: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00015317
CAS číslo: 17499-68-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR