Tisk…

3-Chloro-5-(trifluoromethyl)phenol

Dodavatel: Apollo Scientific
Nebezpečí

PC8004-1G PC8004-25G PC8004-5G
APOSPC8004-1GEA 2070 CZK
APOSPC8004-1G APOSPC8004-25G APOSPC8004-5G
3-Chloro-5-(trifluoromethyl)phenol
3-Chloro-5-(trifluoromethyl)phenol
Vzorec: C₇H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04115852
CAS číslo: 570391-18-3
UN: 3265
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR