Tisk…

Tris(perfluorononyl)-s-triazine 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: SpectraSynQ THT-1485

PC7912-1G
APOSPC7912-1GEA 19350 CZK
APOSPC7912-1G
Tris(perfluorononyl)-s-triazine 95%
Tris(perfluorononyl)-s-triazine

Mass Spectrometry standard

Vzorec: C₃₀F₅₇N₃
MW: 1485.25 g/mol
Bod tání: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00042440
CAS číslo: 57101-59-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR