Tisk…

Methyl perfluoro-3,6,9-trioxatridecanoate

Dodavatel: Apollo Scientific
Varování

PC7879-25G PC7879-5G
APOSPC7879-25GEA 8100 CZK
APOSPC7879-25G APOSPC7879-5G
Methyl perfluoro-3,6,9-trioxatridecanoate
Methyl perfluoro-3,6,9-trioxatridecanoate
Vzorec: C₁₁H₃F₁₉O₅
Bod varu: 73 °C (10 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03094451
CAS číslo: 330562-42-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR