Tisk…

(2,4,5-Trifluorophenyl)acetic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC7818F-100G PC7818F-25G PC7818F-5G PC7818F-500G
APOSPC7818F-100GEA 705 CZK
APOSPC7818F-100G APOSPC7818F-25G APOSPC7818F-5G APOSPC7818F-500G
(2,4,5-Trifluorophenyl)acetic acid 97%
(2,4,5-Trifluorophenyl)acetic acid
Vzorec: (C₆H₂F₃)CH₂COOH
Bod tání: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00082479
CAS číslo: 209995-38-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR