Tisk…

5-Bromo-2-fluorobenzenesulphonyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC7725-5G PC7725-1G PC7725-25G
APOSPC7725-5GEA 827 CZK
APOSPC7725-5G APOSPC7725-1G APOSPC7725-25G
5-Bromo-2-fluorobenzenesulphonyl chloride 98%
5-Bromo-2-fluorobenzenesulphonyl chloride
Vzorec: C₆H₃BrClFO₂S
Bod tání: 61…66 °C
Storage Temperature: Ambient
MDL Number: MFCD09763690
CAS číslo: 339370-40-0
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR