Tisk…

3-(Trifluoromethyl)-L-phenylalanine 97%

Dodavatel: Apollo Scientific

PC7714-1G PC7714-5G
APOSPC7714-1GEA 7010 CZK
APOSPC7714-1G APOSPC7714-5G
3-(Trifluoromethyl)-L-phenylalanine 97%
3-(Trifluoromethyl)-L-phenylalanine
Vzorec: C₁₀H₁₀F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00044945
CAS číslo: 14464-68-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR